Learn More
National Safety Apparel™ FR Hairnet
SDP
Catalog No. 17355204 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
17-355-204 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. 17-355-204 Supplier National Safety Apparel™ Supplier No. H01NYHN
This item is not returnable. View return policy
FR hairnet designed for comfort and flexibility.
  • Soft and breathable FR fabric
  • Designed for comfort and flexibility
  • Elastic fit for comfort
  • FR fabric will not melt, drip, or ignite
  • Full coverage
Specifications
Size Universal
Material Nomex Knit Fabric
Closure Type Pull-over
Disposable Reusable
Arc Flash Rating 2.4 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Quantity 1 Ea.
For Use With (Application) Food Processing, Manufacturing
Resistance Flame
Hood Type None
Style Hairnet