Learn More
MSC Viking Rain Suit, Flame-Resistant, Polyester and PVC, Large, 3-Piece
Catalog No. 5000687044
Encompass
This 3-piece rain suit provides reliable protection and comfort in demanding work environments. Constructed from 0.35 mm thick FR-treated PVC/polyester/PVC sandwich material, the suit offers robust chemical resistance and waterproof, windproof performance. It is designed to meet safety requirements for work sites where flame resistance is necessary.
  • Compliant with CSA Z96-09 Class 1, Level FR standards
  • Features 2 in. yellow prism reflective tape for high visibility
  • Micro-welded seams for enhanced durability
  • Includes a detachable hood and cotton corduroy jacket collar
  • Adjustable features include wrist cuffs, boot cuffs, and a bib pant waist panel
  • Equipped with underarm jacket vents and ESR nylon snap closure systems
  • Suitable for food and fish processing due to oil-resistant sandwich fabric construction
Catalog No. 50-006-87044 Supplier MSC Supplier No. 05272067
This 3-piece rain suit provides reliable protection and comfort in demanding work environments. Constructed from 0.35 mm thick FR-treated PVC/polyester/PVC sandwich material, the suit offers robust chemical resistance and waterproof, windproof performance. It is designed to meet safety requirements for work sites where flame resistance is necessary.
  • Compliant with CSA Z96-09 Class 1, Level FR standards
  • Features 2 in. yellow prism reflective tape for high visibility
  • Micro-welded seams for enhanced durability
  • Includes a detachable hood and cotton corduroy jacket collar
  • Adjustable features include wrist cuffs, boot cuffs, and a bib pant waist panel
  • Equipped with underarm jacket vents and ESR nylon snap closure systems
  • Suitable for food and fish processing due to oil-resistant sandwich fabric construction