Learn More
Hach Company Titration Application Pack Amperometric: Free & Total Chlorine, Chlorine Dioxide, Chlorite, & Sulfite (Autocat)
Catalog No. 501997512 Shop All Hach Company Products
Encompass
Application Pack for the determination of free & total Chlorine, Chlorine Dioxide, Chlorite and Sulphite (Autocat). Suited for Titralab AT1000 Series, Automatic Titrator Models AT1122/AT1222.
Catalog No. 50-199-7512 Supplier Hach Company Supplier No. AP0007.AT1122
Application Pack for the determination of free & total Chlorine, Chlorine Dioxide, Chlorite and Sulphite (Autocat). Suited for Titralab AT1000 Series, Automatic Titrator Models AT1122/AT1222.