Learn More
Medchemexpress LLC FR054 | 35954-65-5 | 99.5% | 329.30 g/mol | C14H19NO8 | 10MG
SDP
Catalog No. 5000235650
Encompass_Preferred
FR054 is a small-molecule inhibitor of the hexosamine biosynthetic pathway enzyme PGM3 used in research to study glycosylation and cancer cell metabolism. It induces early proliferation arrest, endoplasmic reticulum stress, and ROS-dependent apoptotic cell death in breast cancer cells, reduces N- and O-glycosylation levels, and has shown tumor growth suppression in xenograft models.
  • Inhibits PGM3, targeting hexosamine biosynthesis.
  • Induces proliferation arrest and ROS-dependent apoptosis in cancer cells.
  • Reduces N- and O-glycosylation levels in cell models.
  • Shows tumor growth suppression in xenograft studies.
  • Offered with high reported purity for research use.
  • Available as solid forms and as a 10 mM solution in DMSO.
Catalog No. 50-002-35650 Supplier Medchemexpress LLC Supplier No. HY124909A10MG
FR054 is a small-molecule inhibitor of the hexosamine biosynthetic pathway enzyme PGM3 used in research to study glycosylation and cancer cell metabolism. It induces early proliferation arrest, endoplasmic reticulum stress, and ROS-dependent apoptotic cell death in breast cancer cells, reduces N- and O-glycosylation levels, and has shown tumor growth suppression in xenograft models.
  • Inhibits PGM3, targeting hexosamine biosynthesis.
  • Induces proliferation arrest and ROS-dependent apoptosis in cancer cells.
  • Reduces N- and O-glycosylation levels in cell models.
  • Shows tumor growth suppression in xenograft studies.
  • Offered with high reported purity for research use.
  • Available as solid forms and as a 10 mM solution in DMSO.