Learn More
Medchemexpress LLC Fr054 | 35954-65-5 | 99.5% | 329.30 g·mol⁻¹ | C14H19NO8 | 50MG
SDP
Catalog No. 5000237332
Encompass_Preferred
FR054 is a small-molecule research inhibitor of the hexosamine biosynthetic pathway enzyme phosphoglucomutase 3 (PGM3). It alters protein glycosylation, induces endoplasmic reticulum stress and reactive oxygen species-dependent apoptosis in breast cancer cells, and has been shown to suppress tumor growth in MDA-MB-231 xenograft models. Supplied as a high-purity oil for biochemical and cell-based studies.
  • Inhibits PGM3 in the hexosamine biosynthetic pathway.
  • Shows anti-breast cancer activity in cell and xenograft studies.
  • Modulates N- and O-linked protein glycosylation.
  • Induces ER stress and ROS-dependent apoptotic cell death.
  • High purity suitable for research applications.
  • Recommended cold storage to maintain stability.
Catalog No. 50-002-37332 Supplier Medchemexpress LLC Supplier No. HY124909A50MG
Add to Cart
Add to Cart
FR054 is a small-molecule research inhibitor of the hexosamine biosynthetic pathway enzyme phosphoglucomutase 3 (PGM3). It alters protein glycosylation, induces endoplasmic reticulum stress and reactive oxygen species-dependent apoptosis in breast cancer cells, and has been shown to suppress tumor growth in MDA-MB-231 xenograft models. Supplied as a high-purity oil for biochemical and cell-based studies.
  • Inhibits PGM3 in the hexosamine biosynthetic pathway.
  • Shows anti-breast cancer activity in cell and xenograft studies.
  • Modulates N- and O-linked protein glycosylation.
  • Induces ER stress and ROS-dependent apoptotic cell death.
  • High purity suitable for research applications.
  • Recommended cold storage to maintain stability.