Learn More
Cayman Chemical FR122047 (hydrochloride), Purity: ≥98%, CAS: 130717-51-0, 1 mg
SDP
Catalog No. 502889497
Encompass_Preferred
A selective COX-1 inhibitor (IC50 = 0.028 µM for the human enzyme); selective for COX-1 over COX-2 (IC50 = 65 µM); inhibits arachidonic acid, collagen, and ADP-induced platelet aggregation in human platelet-rich plasma (IC50 = 180-200 nM); unlike aspirin, it does not induce gastric damage at 100 mg/kg, p.o.; has an anti-inflammatory effect in a rat models of collagen-induced arthritis
Catalog No. 50-288-9497 Supplier Cayman Chemical Supplier No. 100391 MG
A selective COX-1 inhibitor (IC50 = 0.028 µM for the human enzyme); selective for COX-1 over COX-2 (IC50 = 65 µM); inhibits arachidonic acid, collagen, and ADP-induced platelet aggregation in human platelet-rich plasma (IC50 = 180-200 nM); unlike aspirin, it does not induce gastric damage at 100 mg/kg, p.o.; has an anti-inflammatory effect in a rat models of collagen-induced arthritis