Learn More
Cayman Chemical FR179642hydrate 10mg
SDP
Catalog No. 502990546
Encompass_Preferred
An intermediate in the synthesis of FK463
Catalog No. 50-299-0546 Supplier Cayman Chemical Supplier No. 2964710 MG
An intermediate in the synthesis of FK463