Learn More
Medchemexpress LLC FR194738 | 204067-52-7 | 99.8% | 476.11 | C27H38ClNO2S | 25 MG
SDP
Catalog No. 5000213921
Encompass_Preferred
FR194738 is a small-molecule squalene epoxidase inhibitor for preclinical research. It inhibits squalene epoxidase activity in HepG2 cell homogenates (IC50 = 9.8 nM) and is provided with analytical characterization suitable for biochemical and cellular studies.
  • Potent squalene epoxidase inhibitor (IC50 = 9.8 nM).
  • Suitable for biochemical and cell-based assays.
  • High purity appropriate for analytical applications (~99.8%).
  • Characterized by CAS number 204067-52-7 and molecular weight 476.11.
  • Available in multiple vial sizes, including 25 mg.
  • Supplied with supporting documentation: datasheet, COA, SDS, H NMR, LCMS.
Catalog No. 50-002-13921 Supplier Medchemexpress LLC Supplier No. HY100303A25MG
FR194738 is a small-molecule squalene epoxidase inhibitor for preclinical research. It inhibits squalene epoxidase activity in HepG2 cell homogenates (IC50 = 9.8 nM) and is provided with analytical characterization suitable for biochemical and cellular studies.
  • Potent squalene epoxidase inhibitor (IC50 = 9.8 nM).
  • Suitable for biochemical and cell-based assays.
  • High purity appropriate for analytical applications (~99.8%).
  • Characterized by CAS number 204067-52-7 and molecular weight 476.11.
  • Available in multiple vial sizes, including 25 mg.
  • Supplied with supporting documentation: datasheet, COA, SDS, H NMR, LCMS.