Learn More
Medchemexpress LLC FR194738 | 204067-52-7 | 99.8% | 476.11 | C27H38ClNO2S | 50 MG
SDP
Catalog No. 5000214478
Encompass_Preferred
FR194738 is a potent squalene epoxidase inhibitor used in biochemical and cell-based research to block cholesterol biosynthesis. It inhibits squalene epoxidase in HepG2 cell homogenates with an IC50 of 9.8 nM and reduces cholesterol formation in intact HepG2 cells (IC50s 4.9 nM and 8.0 nM). Supplied as a solid for research use (CAS 204067-52-7); solubility and storage guidance are provided.
Catalog No. 50-002-14478 Supplier Medchemexpress LLC Supplier No. HY100303A50MG
FR194738 is a potent squalene epoxidase inhibitor used in biochemical and cell-based research to block cholesterol biosynthesis. It inhibits squalene epoxidase in HepG2 cell homogenates with an IC50 of 9.8 nM and reduces cholesterol formation in intact HepG2 cells (IC50s 4.9 nM and 8.0 nM). Supplied as a solid for research use (CAS 204067-52-7); solubility and storage guidance are provided.