Learn More
Cayman Chemical FR900098 (sodium salt), Purity: ≥95%, CAS: 73226-73-0, 5 mg
SDP
Catalog No. 502888834
Encompass_Preferred
A derivative of fosmidomycin that has antimalarial activity; inhibits DOXP reductoisomerase in vitro in a dose-dependent manner; exhibits in vitro antimalarial activity against three P. falciparum strains (IC50s = 170, 170, and 90 nM for HB3, A2, and Dd2 strains, respectively); FR900098 (>10 mg/kg, i.p.) eradicated P. vinckei parasites within four days in mice
Catalog No. 50-288-8834 Supplier Cayman Chemical Supplier No. 100126815 MG
A derivative of fosmidomycin that has antimalarial activity; inhibits DOXP reductoisomerase in vitro in a dose-dependent manner; exhibits in vitro antimalarial activity against three P. falciparum strains (IC50s = 170, 170, and 90 nM for HB3, A2, and Dd2 strains, respectively); FR900098 (>10 mg/kg, i.p.) eradicated P. vinckei parasites within four days in mice