Learn More
Medchemexpress LLC HY-16212 1mg , FR901464 CAS:146478-72-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY162121MG
Encompass_Preferred
HY-16212 1mg FR901464 CAS: 146478-72-0 FR901464 is a potent spliceosome inhibitor with prominent anti-tumor and anti-cancer effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0969