Learn More
Cole-Parmer General Tools 147 Digital Stainless Steel Caliper, 0 to 6" with Fractions," or Milimeters UOM
Catalog No. NC9032922 Shop All Cole-Parmer Products
Encompass_Preferred
Direct inch/metric/fraction conversion; measure inside, outside, depth, step; friction thumb roller.
Catalog No. NC9032922 Supplier Cole-Parmer Supplier No. 9715214
Direct inch/metric/fraction conversion; measure inside, outside, depth, step; friction thumb roller.