Learn More
Medchemexpress LLC Fractalkine/CX3CL1 2ug
SDP
Catalog No. 5000182107
Encompass_Preferred
The Fractalkine/CX3CL1 protein is a multifunctional chemokine that binds to CX3CR1 and the integrins ITGAV ITGB3 and ITGA4 ITGB1 It regulates immune responses inflammation cell adhesion and chemotaxis Fractalkine/CX3CL1 Protein Mouse (HEK293 His) is the recombinant mouse-derived Fractalkine/CX3CL1 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82107 Supplier Medchemexpress LLC Supplier No. HYP726862UG
The Fractalkine/CX3CL1 protein is a multifunctional chemokine that binds to CX3CR1 and the integrins ITGAV ITGB3 and ITGA4 ITGB1 It regulates immune responses inflammation cell adhesion and chemotaxis Fractalkine/CX3CL1 Protein Mouse (HEK293 His) is the recombinant mouse-derived Fractalkine/CX3CL1 protein expressed by HEK293 with C-6 His labeled tag