Learn More
Medchemexpress LLC Fractalkine/CX3CL1 50ug | 50ug
SDP
Catalog No. 5000187455
Encompass_Preferred
The Fractalkine/CX3CL1 protein is a multifunctional chemokine that binds to CX3CR1 and the integrins ITGAV ITGB3 and ITGA4 ITGB1 It regulates immune responses inflammation cell adhesion and chemotaxis Fractalkine/CX3CL1 Protein Mouse (Biotinylated HEK293 His-Avi) is the recombinant mouse-derived Fractalkine/CX3CL1 protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87455 Supplier Medchemexpress LLC Supplier No. HYP7811350UG
The Fractalkine/CX3CL1 protein is a multifunctional chemokine that binds to CX3CR1 and the integrins ITGAV ITGB3 and ITGA4 ITGB1 It regulates immune responses inflammation cell adhesion and chemotaxis Fractalkine/CX3CL1 Protein Mouse (Biotinylated HEK293 His-Avi) is the recombinant mouse-derived Fractalkine/CX3CL1 protein expressed by HEK293 with C-Avi C-His labeled tag