Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fraction V Bovine Albumin, pH 7

Catalog No. AAJ1085718
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ1085718 50 g
AAJ1085722 100 g
AAJ1085736 500 g
AAJ10857A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ1085718 Supplier Thermo Scientific Supplier No. J1085718
Add to Cart

Heat-Shock fractionated

  • Source: Bovine blood
  • Form: Lyophilized powder
  • Assay (electrophoresis): ≥98.0%
  • Protein: ≥96.0%
  • pH (2%, H2O): 6.5 - 7.5
  • Loss On Drying: ≤5.0%
  • Residue on Ignition: ≤2.0%
  • CAS: 9048-46-8
TRUSTED_SUSTAINABILITY

Specifications

pH Range 6.5 to 7.5
Quantity 50 g
Source Bovine blood
Storage Requirements 4°C
Packaging Plastic Bottle
Product Type Fraction V Bovine Albumin
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.