Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Fractional Distillation
GREENER_CHOICESCI_ED

Catalog No. S42859 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S42859 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Methanol and Phthalates, which are known to the State of California to cause and reproductive harm.
Catalog No. S42859 Supplier Innovating Science™ Supplier No. IS2521
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Methanol and Phthalates, which are known to the State of California to cause and reproductive harm.

Ideal for studying topics such as boiling and condensation points, conservation of matter. Innovating Science™ Fractional Distillation is a fractional distillation experiment to simulate the fractionation of crude oil.

  • Crude oil is a complex mixture of many useful hydrocarbons such as gasoline, kerosene, butane, and motor oil
  • In order to be of use, these hydrocarbons must first be separated from each other
  • This Fractional Distillation lab activity allows students to complete a fractional distillation experiment that simulates the fractionation of crude oil
  • Topics such as boiling and condensation points, conservation of matter, and forces between molecules are covered as they factor into how a distillation is performed
  • This kit contains enough material for 15 groups
  • Teacher's Manual and Student Study Guide copymasters are included
  • Kit Includes:
    • Simulated Crude Oil (450 mL)
    • 15 Rubber Stoppers
    • 15 pieces Tubing
    • 30 Disposable Cups

Specifications

Certifications/Compliance Product SDSs
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Science Education
Includes 15 Two-hole Stoppers for 250 mL Erlenmeyer Flask 15 pcs Plastic Tubing 450 mL Simulated Crude Oil 30 Graduated Medicine Cups
Quantity 1 Kit
Weight (English) 3.35 lb.
Dimensions (L x W x H) 9 x 9 x 9 in.
Class Size 15 Lab Groups
Product Type General Chemistry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.