Learn More
Grainger Fractional Tap and Die Set 17 pcs.
Catalog No. 502752551
Encompass_Preferred
The West Indigo 141H77 17-piece fractional tap and die set includes taps and dies for cutting and threading fractional-sized holes and bolts from 1/4" to 1/2" in 1/64" increments. The carbon steel taps and dies have a black oxide finish for durability and rust resistance. The set comes in a plastic blow-molded case and includes a tap wrench, die stock, screwdriver, and pitch gauge for ease of use. This tap and die set is ideal for home DIY projects and light repair work requiring fractional inch thread sizes.
Catalog No. 50-275-2551 Supplier Grainger Supplier No. 141H77
The West Indigo 141H77 17-piece fractional tap and die set includes taps and dies for cutting and threading fractional-sized holes and bolts from 1/4" to 1/2" in 1/64" increments. The carbon steel taps and dies have a black oxide finish for durability and rust resistance. The set comes in a plastic blow-molded case and includes a tap wrench, die stock, screwdriver, and pitch gauge for ease of use. This tap and die set is ideal for home DIY projects and light repair work requiring fractional inch thread sizes.