Learn More
Grainger Fractional Tap/Hexagon Die Set 14 pcs.
Catalog No. 502752490
Encompass_Preferred
The DEWALT Fractional Tap and Hexagon Die Set is a high-quality tap and die set for cutting and threading fractional-sized threads. The set includes 21 taps and 21 dies with sizes ranging from 4-40 to 1/4-20. The taps and dies are made from hardened alloy steel for durability and have a black oxide finish to prevent rust. The set comes organized in a molded case for convenient storage and portability. This is an essential tool for any metalworker looking to cut internal and external threads on nuts, bolts, pipes, and other hardware.
Catalog No. 50-275-2490 Supplier Grainger Supplier No. 141H78
The DEWALT Fractional Tap and Hexagon Die Set is a high-quality tap and die set for cutting and threading fractional-sized threads. The set includes 21 taps and 21 dies with sizes ranging from 4-40 to 1/4-20. The taps and dies are made from hardened alloy steel for durability and have a black oxide finish to prevent rust. The set comes organized in a molded case for convenient storage and portability. This is an essential tool for any metalworker looking to cut internal and external threads on nuts, bolts, pipes, and other hardware.