Learn More
COANN TECHNOLOGIES LLC Protein/Proteoform Fractionation Column, 100 um ID x 50 cm L x 365 um OD, 3 um C2 (300A)
Catalog No. NC3781362
Encompass
Flow Rate (nL/min)- 450 ± 150. Operation Pressure (psi)- <8000. Column Feature- Separate
Catalog No. NC3781362 Supplier COANN TECHNOLOGIES LLC Supplier No. HPF10005003002SWF
Flow Rate (nL/min)- 450 ± 150. Operation Pressure (psi)- <8000. Column Feature- Separate