Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Fractions and Decimals Curriculum Mastery Flip Chart Set
SCI_ED

Catalog No. S24119 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
3 to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S24119 3 to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24119 Supplier NewPath Learning™ Supplier No. 333005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Set includes

  • Place Value - Understanding Numbers
  • Understanding Fractions
  • All About Fractions
  • All About Decimals
  • Adding & Subtracting Decimals
  • Fraction Concepts
  • Adding & Subtracting Fractions
  • Multiplying & Dividing Decimals
  • Multiplying & Dividing Fractions
  • Operations with Mixed Numbers

Activity Guide featuring questions and exercises.

  • Flip chart mounted on sturdy easel
  • Features 10 double-sided laminated 12 x 18 in. charts
  • Side 1 displays colorful, graphic overview of topic
  • Side 2 features Write-On/Wipe-Off activity Chart features black-line copy-masters, questions, labeling exercises, vocabulary review, etc

Specifications

Grade Level Elementary
Grade 3 to 5
Format Chart
Product Type Fractions and Decimals Curriculum Mastery Flip Chart Set
Includes
1. Place Value - Understanding Numbers
2. Understanding Fractions
3. All About Fractions
4. All About Decimals
5. Adding & Subtracting Decimals
6. Fraction Concepts
7. Adding & Subtracting Fractions
8. Multiplying & Dividing Decimals
9. Multiplying & Dividing Fractions
10. Operations with Mixed Numbers
Certifications Academic standard: California, Florida, National, New York, Texas
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.