Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fradafiban hydrochloride | 00-00-0 | MFCD00919205 | 100.0% | 403.86 g·mol⁻¹ | C20H22ClN3O4 | 1 MG
SDP

Catalog No. 5000245722
Encompass_Preferred

Fradafiban hydrochloride is a nonpeptide platelet glycoprotein IIb/IIIa antagonist that binds the human GP IIb/IIIa complex (Kd = 148 nM) and inhibits platelet aggregation in vitro. It is a nonpeptide mimetic of the arginine-glycine-aspartic acid (RGD) recognition sequence, supplied as a white to off-white solid for research use.

  • Binds human platelet GP IIb/IIIa with a reported Kd of 148 nM.
  • Inhibits platelet aggregation in vitro.
  • Nonpeptide RGD mimetic, suitable for mechanistic studies.
  • High purity (reported 99.96%).
  • White to off-white solid, convenient for handling and storage.
  • Available in small research sizes (1 mg, 5 mg, 10 mg, 50 mg, 100 mg).
  • Molecular weight 403.86 g·mol⁻¹; molecular formula C20H22ClN3O4.

Catalog No. 50-002-45722 Supplier Medchemexpress LLC Supplier No. HY101720A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fradafiban hydrochloride is a nonpeptide platelet glycoprotein IIb/IIIa antagonist that binds the human GP IIb/IIIa complex (Kd = 148 nM) and inhibits platelet aggregation in vitro. It is a nonpeptide mimetic of the arginine-glycine-aspartic acid (RGD) recognition sequence, supplied as a white to off-white solid for research use.

  • Binds human platelet GP IIb/IIIa with a reported Kd of 148 nM.
  • Inhibits platelet aggregation in vitro.
  • Nonpeptide RGD mimetic, suitable for mechanistic studies.
  • High purity (reported 99.96%).
  • White to off-white solid, convenient for handling and storage.
  • Available in small research sizes (1 mg, 5 mg, 10 mg, 50 mg, 100 mg).
  • Molecular weight 403.86 g·mol⁻¹; molecular formula C20H22ClN3O4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.