Learn More
Jackson Immuno Research Labs AffiniPure F(ab')2 Fragment Donkey Anti-Mouse IgM, u chain specific, 1 mg
SDP
Catalog No. NC1656705
Encompass_Preferred
F(ab')2 fragment antibodies are generated by pepsin digestion of whole IgG antibodies to remove most of the Fc region while leaving some of the hinge region. F(ab')2 fragments have two antigen-binding Fab portions linked together by disulfide bonds and therefore they are divalent. The average molecular weight is about 110 kDa. They are used for specific applications, such as to avoid binding of secondary antibodies to live cells with Fc receptors or to Protein A or Protein G.
Catalog No. NC1656705 Supplier Jackson Immuno Research Labs Supplier No. 715006020
F(ab')2 fragment antibodies are generated by pepsin digestion of whole IgG antibodies to remove most of the Fc region while leaving some of the hinge region. F(ab')2 fragments have two antigen-binding Fab portions linked together by disulfide bonds and therefore they are divalent. The average molecular weight is about 110 kDa. They are used for specific applications, such as to avoid binding of secondary antibodies to live cells with Fc receptors or to Protein A or Protein G.