Learn More
Medchemexpress LLC Frakefamide TFA | 00-00-0 | 99.2% | 677.64 g·mol⁻¹ | C32H35F4N5O7 | 5 MG
SDP
Catalog No. 5000243666
Encompass_Preferred
Frakefamide TFA is a peptide analgesic supplied as the trifluoroacetate salt that acts as a peripherally selective μ-opioid receptor agonist. It has limited ability to cross the blood-brain barrier, making it useful for research into peripheral analgesia and opioid receptor pharmacology. The product is provided in small solid quantities and as prepared DMSO solutions, with manufacturer-specified purity and molecular data.
Catalog No. 50-002-43666 Supplier Medchemexpress LLC Supplier No. HY106147B5MG
Frakefamide TFA is a peptide analgesic supplied as the trifluoroacetate salt that acts as a peripherally selective μ-opioid receptor agonist. It has limited ability to cross the blood-brain barrier, making it useful for research into peripheral analgesia and opioid receptor pharmacology. The product is provided in small solid quantities and as prepared DMSO solutions, with manufacturer-specified purity and molecular data.