Learn More
Medchemexpress LLC Frakefamide TFA 10mM 1mL | 10MM 1ML
SDP
Catalog No. 5000242939
Encompass_Preferred
Frakefamide TFA is a potent analgesic that acts as a peripheral active -selective receptor agonist Frakefamide is unable to penetrate the blood-brain-barrier and enter the central nervous system[1 [2
Catalog No. 50-002-42939 Supplier Medchemexpress LLC Supplier No. HY106147B10MM 1ML
Frakefamide TFA is a potent analgesic that acts as a peripheral active -selective receptor agonist Frakefamide is unable to penetrate the blood-brain-barrier and enter the central nervous system[1 [2