Learn More
Medchemexpress LLC Frakefamide TFA 25mg | 25MG
SDP
Catalog No. 5000243733
Encompass_Preferred
Frakefamide TFA is a potent analgesic that acts as a peripheral active -selective receptor agonist Frakefamide is unable to penetrate the blood-brain-barrier and enter the central nervous system[1 [2
Catalog No. 50-002-43733 Supplier Medchemexpress LLC Supplier No. HY106147B25MG
Frakefamide TFA is a potent analgesic that acts as a peripheral active -selective receptor agonist Frakefamide is unable to penetrate the blood-brain-barrier and enter the central nervous system[1 [2