Learn More
OHAUS™ Frame Connector Clamp
Catalog No. 01922036 Shop All OHAUS Products
Change view
Click to view available options
Model:
CLC-HOOKCZ
Catalog No. Model
01-922-036 CLC-HOOKCZ
1 options
Catalog No. 01-922-036 Supplier OHAUS™ Supplier No. 30392258
Add to Cart
Add to Cart
LabJaws ensures secure support for holding your labware and apparatus
  • Lab-frame kits include all components necessary to build a lab-frame
  • LabJaws offers the largest selection of Lab-Frame Kits, Lab-Lifts, Rods, Support Stands & Connectors in the industry to safely hold labware.
  • Whether you need a standard lab-frame or need to create a unique set-up, we have options to fit any space or application.
  • Dependable connectors, secure rods, and clamps to meet your requirements.
  • Five unique holders available in a variety of sizes and materials to accommodate different rods up to 24 mm in diameter
TRUSTED_SUSTAINABILITY
Specifications
Description Grip Range: 0 in - 0.51 in (0 mm - 13 mm)
For Use With (Application) Chromatography, Distillation, Titrations, Electrolysis, Chemistry, Research and Development, Quality Control, Ring Stands, Water Baths, Cleanroom Applications, Acid Fume Hood Applications
Includes Clamp, Connector, Hook, Thumbscrew adjustment
Model CLC-HOOKCZ
Shape Connection Angle: Fixed 90°
Type Frame connector CLC-HOOKCZ 0-13 mm
Material Metal