Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Frames and Caps for Immuno Standard Modules

Catalog No. 12565131 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Clear
White
Sterility:
Non-sterile
Sterile
Product Type:
Frame
Strip Cap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Product Type
12-565-131 White Non-sterile Frame
12-565-260 Clear Non-sterile Strip Cap
12-565-262 Clear Sterile Strip Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-565-131 Supplier Thermo Scientific™ Supplier No. 460348
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easily insert and remove strip plates with solid modules by utilizing these frames and caps for immuno standard modules.

Easily insert and remove strip plates with solid modules by utilizing Thermo Scientific™ Frames and Caps for Immuno Standard Modules.

  • Polyethylene, stackable frame is marked for lid orientation
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Frame
For Use With (Equipment) Nunc Modules
Material Polyethylene
Sterility Non-sterile
No. per Case 60
No. per Pack 5
Plate Compatibility Immuno Standard Modules
Quantity 60/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.