Learn More
Thermo Scientific™ Frames and Caps for Immuno Standard Modules
GREENER_CHOICE
Catalog No. 12565260
Change view
Click to view available options
Color:
Clear
White
Sterility:
Non-sterile
Sterile
Product Type:
Frame
Strip Cap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Product Type
12-565-260 Clear Non-sterile Strip Cap
12-565-262 Clear Sterile Strip Cap
12-565-131 White Non-sterile Frame
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-565-260 Supplier Thermo Scientific™ Supplier No. 430082
Add to Cart
Add to Cart
Easily insert and remove strip plates with solid modules by utilizing these frames and caps for immuno standard modules.
Easily insert and remove strip plates with solid modules by utilizing Thermo Scientific™ Frames and Caps for Immuno Standard Modules.
  • Polyethylene, stackable frame is marked for lid orientation
TRUSTED_SUSTAINABILITY
Specifications
Format Strip Plate
Array 1 x 8
Packaging 180 Cases, 60 Pk.
Color Clear
Product Type Strip Cap
Material Polyethylene
No. of Wells 8
Sterility Non-sterile
No. per Case 180
No. per Pack 60
Plate Compatibility Immuno Standard Modules
Quantity 80/Cs.