Learn More
Thermo Scientific™ Frames and Caps for Immuno Standard Modules
GREENER_CHOICE
Catalog No. 12565262 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Clear
White
Sterility:
Non-sterile
Sterile
Product Type:
Frame
Strip Cap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Product Type
12-565-262 Clear Sterile Strip Cap
12-565-260 Clear Non-sterile Strip Cap
12-565-131 White Non-sterile Frame
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 12-565-262 Supplier Thermo Scientific™ Supplier No. 430805
  • $223.00
    $20.77 / Case of 120
    Save $202.23
    91% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Easily insert and remove strip plates with solid modules by utilizing these frames and caps for immuno standard modules.
Easily insert and remove strip plates with solid modules by utilizing Thermo Scientific™ Frames and Caps for Immuno Standard Modules.
  • Polyethylene, stackable frame is marked for lid orientation
TRUSTED_SUSTAINABILITY
Specifications
Format Strip Plate
Array 1 x 8
Packaging 2 Pk.
Color Clear
Product Type Strip Cap
Material Polyethylene
No. of Wells 8
Sterility Sterile
No. per Case 120
No. per Pack 2
Plate Compatibility Immuno Standard Modules
Quantity 120/Cs.
Show More Show Less
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?
Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with LockWell modules?
Due to the locking feature design of the LockWell modules, the 8-Well Strip Caps for Immuno Standard Modules are not compatible with, and will not seal, the LockWell Modules.