Learn More
Labsource Inc FrameStar 384 Well Skirted PCR Plate, High-Performance for PCR, Ultra-Low DNA Binding, 50 Plates/Pack
SDP
Catalog No. NC2946449
Encompass
The FrameStar 384 Well Skirted PCR Plate is designed for high-throughput PCR and qPCR applications. Its thin-walled polypropylene wells optimize thermal transfer, while the rigid polycarbonate frame prevents warping during rapid cycling, ensuring enhanced seal integrity. Features include ultra-low DNA binding properties, making it ideal for sensitive experiments. Compatible with most 384-well block instruments, this robust plate guarantees consistency and minimal sample evaporation during assays.
Catalog No. NC2946449 Supplier Labsource Inc Supplier No. P97109
Add to Cart
Add to Cart
The FrameStar 384 Well Skirted PCR Plate is designed for high-throughput PCR and qPCR applications. Its thin-walled polypropylene wells optimize thermal transfer, while the rigid polycarbonate frame prevents warping during rapid cycling, ensuring enhanced seal integrity. Features include ultra-low DNA binding properties, making it ideal for sensitive experiments. Compatible with most 384-well block instruments, this robust plate guarantees consistency and minimal sample evaporation during assays.