Learn More
World Precision Instrument FrameWorks V-Base Kit - Large
SDP
Catalog No. 502521298
Encompass
These high quality bases are made of stainless steel and polymer that resists organic solvents and corrosion. They can be easily assembled to make a stand-alone setup for student laboratory equipment or to make a complicated frame for research labs. Use these components to custom build your own stands for laboratory glassware, manipulator stands or any other laboratory application you can envision.
Catalog No. 50-252-1298 Supplier World Precision Instrument Supplier No. 503208
These high quality bases are made of stainless steel and polymer that resists organic solvents and corrosion. They can be easily assembled to make a stand-alone setup for student laboratory equipment or to make a complicated frame for research labs. Use these components to custom build your own stands for laboratory glassware, manipulator stands or any other laboratory application you can envision.