Learn More
Medchemexpress LLC Framycetin sulfate | 4146-30-9 | 98.0% | 908.88 | 500 MG
SDP
Catalog No. 5000386984
Encompass_Preferred
Framycetin sulfate is an aminoglycoside antibiotic that acts as a potent inhibitor of RNase P cleavage activity. It competes for specific divalent metal ion binding sites in RNase P RNA and also inhibits hammerhead ribozyme. This compound is a 5″-azido neomycin B precursor and binds to the Drosha site in miR-525. It is used in the treatment of hepatic encephalopathy and infections caused by enteropathogenic E. coli.
  • Potent RNase P cleavage activity inhibitor
  • Inhibits hammerhead ribozyme
  • Binds to the Drosha site in miR-525
  • Effective against hepatic encephalopathy and enteropathogenic E. coli infections
Catalog No. 50-003-86984 Supplier Medchemexpress LLC Supplier No. HY17624A500MG
Framycetin sulfate is an aminoglycoside antibiotic that acts as a potent inhibitor of RNase P cleavage activity. It competes for specific divalent metal ion binding sites in RNase P RNA and also inhibits hammerhead ribozyme. This compound is a 5″-azido neomycin B precursor and binds to the Drosha site in miR-525. It is used in the treatment of hepatic encephalopathy and infections caused by enteropathogenic E. coli.
  • Potent RNase P cleavage activity inhibitor
  • Inhibits hammerhead ribozyme
  • Binds to the Drosha site in miR-525
  • Effective against hepatic encephalopathy and enteropathogenic E. coli infections