Learn More
Selleck Chemical LLC Framycetin sulphate-E0511-25MG
SDP
Catalog No. 503136921
Encompass_Preferred
Framycetin sulfate (Neomycin B sulfate) an aminoglycoside antibiotic is a potent RNase P cleavage activity inhibitor with a Ki of 35 M and competes for specific divalent metal ion binding sites in RNase P RNA also inhibits hammerhead ribozyme with a Ki of 13 5 M
Catalog No. 50-313-6921 Supplier Selleck Chemical LLC Supplier No. E051125MG
Framycetin sulfate (Neomycin B sulfate) an aminoglycoside antibiotic is a potent RNase P cleavage activity inhibitor with a Ki of 35 M and competes for specific divalent metal ion binding sites in RNase P RNA also inhibits hammerhead ribozyme with a Ki of 13 5 M