Learn More
IBL America Francisella Tularensis IgM ELISA
Catalog No. 501538148
Encompass_Preferred
Enzyme immunoassay (ELISA) for the determination of human IgM antibodies directed against the lipopolysaccharide (LPS) of Francisella tularensis in serum or plasma (EDTA, citrate, heparin). For research use only, not for use in diagnostic procedures.Thereaction is based on the specific interaction of antibodies with their corresponding antigen.The test strips of the microtiter plate are coated with specific antigens of the pathogen ofinterest. If antibodies in the sample are present, they bind to the fixed antigen. Asecondary antibody, which has been conjugated with the enzyme alkaline phosphatase,detects and binds to the immune complex. The colourless substrate pnitrophenylphosphate is then converted into the coloured product p-nitrophenol. Thesignal intensity of this reaction product is proportional to the concentration of the analyte inthe sample and is measured photometrically.
Catalog No. 50-153-8148 Supplier IBL America Supplier No. IB05059
Add to Cart
Add to Cart
Enzyme immunoassay (ELISA) for the determination of human IgM antibodies directed against the lipopolysaccharide (LPS) of Francisella tularensis in serum or plasma (EDTA, citrate, heparin). For research use only, not for use in diagnostic procedures.Thereaction is based on the specific interaction of antibodies with their corresponding antigen.The test strips of the microtiter plate are coated with specific antigens of the pathogen ofinterest. If antibodies in the sample are present, they bind to the fixed antigen. Asecondary antibody, which has been conjugated with the enzyme alkaline phosphatase,detects and binds to the immune complex. The colourless substrate pnitrophenylphosphate is then converted into the coloured product p-nitrophenol. Thesignal intensity of this reaction product is proportional to the concentration of the analyte inthe sample and is measured photometrically.