Learn More
Medchemexpress LLC Frangufoline | 19526-09-1 | 98.4% | 1 MG
SDP
Catalog No. 5000234656
Encompass_Preferred
Frangufoline is a 4(14)-type cyclopeptide alkaloid isolated from the root bark of Ziziphus cambodiana Pierre. It is provided as a purified small-molecule research reagent (CAS 19526-09-1) with reported molecular weight 534.69 g/mol and high purity for biochemical studies.
  • 4(14)-type cyclopeptide alkaloid isolated from plant root bark.
  • High purity suitable for biochemical and pharmacological research.
  • Supplied in small quantities for screening and structure-activity studies.
  • Storage: store at 4°C and protect from light; in solvent, keep at -80°C (6 months) or -20°C (1 month).
  • Provided with data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-34656 Supplier Medchemexpress LLC Supplier No. HY1196371MG
Frangufoline is a 4(14)-type cyclopeptide alkaloid isolated from the root bark of Ziziphus cambodiana Pierre. It is provided as a purified small-molecule research reagent (CAS 19526-09-1) with reported molecular weight 534.69 g/mol and high purity for biochemical studies.
  • 4(14)-type cyclopeptide alkaloid isolated from plant root bark.
  • High purity suitable for biochemical and pharmacological research.
  • Supplied in small quantities for screening and structure-activity studies.
  • Storage: store at 4°C and protect from light; in solvent, keep at -80°C (6 months) or -20°C (1 month).
  • Provided with data sheet, certificate of analysis, and safety data sheet.