Learn More
MSC Franklin Fibre-Lamitex Goggle Retainer Clip, Welder's Hat, Orange, 1 ea
Catalog No. 5000767212
Encompass
Franplas Safety Goggle Retainer Clips are designed to securely hold safety goggles on top of hard hats, allowing for quick deployment. They easily snap onto safety helmets, ensuring goggles are readily available for instant use.
  • Thermoformed from high impact, chemical resistant Kydex PVC/Acrylic composite material.
  • Designed for welder's hat style hard hats with a full brim approximately 1½ in all around.
  • Allows for instant access to safety goggles.
Catalog No. 50-007-67212 Supplier MSC Supplier No. 53168126
Franplas Safety Goggle Retainer Clips are designed to securely hold safety goggles on top of hard hats, allowing for quick deployment. They easily snap onto safety helmets, ensuring goggles are readily available for instant use.
  • Thermoformed from high impact, chemical resistant Kydex PVC/Acrylic composite material.
  • Designed for welder's hat style hard hats with a full brim approximately 1½ in all around.
  • Allows for instant access to safety goggles.