Learn More
Cell Biolabs Inc OxiSelect™ Ferric Reducing Antioxidant Power (FRAP) Assay Kit, 200 assays
Catalog No. 501098809
Encompass_Preferred
The Ferric Reducing Antioxidant Power (FRAP) Assay is a simple method for quantifying the antioxidant capacity of a sample. A ferric (Fe3+) ion is reduced to its ferrous (Fe2+) form by electron-donating antioxidants present in the sample. Cell Biolabs' OxiSelect™ Ferric Reducing Antioxidant Power (FRAP) Assay Kit is a quantitative assay for measuring the antioxidant potential within various sample types including serum, plasma, cell or tissue lysates, and food extracts. The kit provides sufficient reagents for 200 assays including standards and unknowns.
Catalog No. 50-109-8809 Supplier Cell Biolabs Inc Supplier No. STA859
The Ferric Reducing Antioxidant Power (FRAP) Assay is a simple method for quantifying the antioxidant capacity of a sample. A ferric (Fe3+) ion is reduced to its ferrous (Fe2+) form by electron-donating antioxidants present in the sample. Cell Biolabs' OxiSelect™ Ferric Reducing Antioxidant Power (FRAP) Assay Kit is a quantitative assay for measuring the antioxidant potential within various sample types including serum, plasma, cell or tissue lysates, and food extracts. The kit provides sufficient reagents for 200 assays including standards and unknowns.