Learn More
Arbor Assays FRAP (Ferric Reducing Antioxidant Power) Detection Kit (2 Plate)
SDP
Catalog No. 502278454
Encompass_Preferred
The DetectX FRAP  (Ferric Reducing Antioxidant Power) Detection Kit quantitatively measures antioxidant status in a variety of samples. The assay measures the antioxidant ability from all species.
This item is not returnable. View return policy
Catalog No. 50-227-8454 Supplier Arbor Assays Supplier No. K043H1
The DetectX FRAP  (Ferric Reducing Antioxidant Power) Detection Kit quantitatively measures antioxidant status in a variety of samples. The assay measures the antioxidant ability from all species.