Learn More
Bioworld Fraser Broth, 500 g
Supplier:  Bioworld 306200261
Encompass_Preferred
Fraser BrothFor the isolation and selective enrichment of Listeria species from food and environmental speciesContains Fraser Supplement SolutionpH: 7.2Working Concentration: 55 g/LDehydrated Appearance: tan, homogeneous powderPrepared Appearance: medium amber brothSoluble in boiling waterComposition:Sodium Chloride Disodium Hydrogen Phosphate Beef Extract Proteose Peptone Pancreatic Digest of Casein Yeast Extract Lithium Chloride Potassium Dihydrogen Phosphate Esculin Fraser Supplement Solution
Catalog No. 50-254-304