Learn More
NEOGEN Fraser Broth Base, 500 g
SDP
Catalog No. 502015345
Encompass_Preferred
Fraser Broth Base is used with Acriflavin, Nalidixic Acid, and Ferric Ammonium Citrate for the selective enrichment of Listeria spp. Fraser Broth Base is not intended for use in the diagnosis of disease or other conditions in humans.Formulated according to MLG Appendix 1.09 (for the ISO 11290 formulation please use NCM0001). Listeria monocytogenes, described first in 1926 by Murray, Webb, and Swann, is an extensive problem in public health and food industries. This organism has the ability to cause human illness and death, particularly in immunocompromised individuals and pregnant women. Epidemiological evidence from outbreaks of listeriosis indicated the principle route of transmission is via the consumption of foodstuffs contaminated with Listeria monocytogenes. Implicated vehicles of transmission include turkey frankfurters, coleslaw, pasteurized milk, Mexican style cheese and pate.For more information visit https://www.neogen.com/solutions/Microbiology/fraser-broth-base
Catalog No. 50-201-5345 Supplier NEOGEN Supplier No. 700003157
Fraser Broth Base is used with Acriflavin, Nalidixic Acid, and Ferric Ammonium Citrate for the selective enrichment of Listeria spp. Fraser Broth Base is not intended for use in the diagnosis of disease or other conditions in humans.Formulated according to MLG Appendix 1.09 (for the ISO 11290 formulation please use NCM0001). Listeria monocytogenes, described first in 1926 by Murray, Webb, and Swann, is an extensive problem in public health and food industries. This organism has the ability to cause human illness and death, particularly in immunocompromised individuals and pregnant women. Epidemiological evidence from outbreaks of listeriosis indicated the principle route of transmission is via the consumption of foodstuffs contaminated with Listeria monocytogenes. Implicated vehicles of transmission include turkey frankfurters, coleslaw, pasteurized milk, Mexican style cheese and pate.For more information visit https://www.neogen.com/solutions/Microbiology/fraser-broth-base