Learn More
Medchemexpress LLC HY-15542A 5mg Medchemexpress, FRAX597 CAS:1286739-19-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15542A5MG
Encompass_Preferred
Medchemexpress, HY-15542A 5mg FRAX597 CAS:1286739-19-2 FRAX597 is a potent group I p21-activated Kinases (PAKs) inhibitor with IC50 of 8, 13 and 19 nM for PAK1, 2 and 3. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0645