Learn More
Medchemexpress LLC HY-N0580 5mg Medchemexpress, Fraxetin CAS:574-84-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN05805MG
Encompass_Preferred
Medchemexpress, HY-N0580 5mg Fraxetin CAS:574-84-5 Fraxetin is isolated from Cortex Fraxini. Fraxetin has antitumor, anti-oxidation effects and anti-inflammory effects. Fraxetin induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0291