Learn More
Cayman Chemical Fraxinellone 10mg
SDP
Catalog No. 502949873
Encompass_Preferred
A liminoid degradant; has antifeedant activity against, and inhibits development of, armyworm larvae when applied to fresh wheat leaves at 5, 10, or 20 mg/ml; reduces LPS-induced nuclear translocation of the NF-κB subunit p65, as well as inhibits LPS-induced production of NO and PGE2 in RAW 264.7 cells; reduces tumor growth in an A549 mouse xenograft model at 30 or 100 mg/kg
Catalog No. 50-294-9873 Supplier Cayman Chemical Supplier No. 3467210 MG
A liminoid degradant; has antifeedant activity against, and inhibits development of, armyworm larvae when applied to fresh wheat leaves at 5, 10, or 20 mg/ml; reduces LPS-induced nuclear translocation of the NF-κB subunit p65, as well as inhibits LPS-induced production of NO and PGE2 in RAW 264.7 cells; reduces tumor growth in an A549 mouse xenograft model at 30 or 100 mg/kg