Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fraxinellone | 28808-62-0 | MFCD11101451 | 100.0% | 232.28 | C14H16O3 | 10 MG
SDP

Catalog No. 5000296304
Encompass_Preferred

Fraxinellone is a naturally occurring limonoid research compound used in biochemical and cellular studies. It is supplied as a high-purity solid (CAS 28808-62-0) with molecular formula C14H16O3 and molecular weight 232.28 g/mol.

  • High purity (99.99%).
  • Solid powder suitable for analytical and biological research.
  • Molecular weight 232.28 g/mol and formula C14H16O3.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes including 10 MG for research use.

Catalog No. 50-002-96304 Supplier Medchemexpress LLC Supplier No. HYN024210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fraxinellone is a naturally occurring limonoid research compound used in biochemical and cellular studies. It is supplied as a high-purity solid (CAS 28808-62-0) with molecular formula C14H16O3 and molecular weight 232.28 g/mol.

  • High purity (99.99%).
  • Solid powder suitable for analytical and biological research.
  • Molecular weight 232.28 g/mol and formula C14H16O3.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes including 10 MG for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.