Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fraxinol | 486-28-2 | MFCD08693526 | 98.8% | 222.19 g·mol⁻¹ | C11H10O5 | 10 MG
SDP

Catalog No. 5000289026
Encompass_Preferred

Fraxinol is a naturally derived coumarin isolated from Lobelia chinensis and used as a cytochrome P450 inhibitor for research applications. It is supplied as a light yellow solid with high reported purity, documented solubility in DMSO, and recommended storage conditions for both solid and solution forms.

  • Purity: 98.82%.
  • CAS number: 486-28-2.
  • Molecular formula: C11H10O5; molecular weight: 222.19 g·mol⁻¹.
  • Biological target: cytochrome P450 enzymes.
  • Solubility: DMSO 25 mg/mL (in vitro); in vivo formulations yield ≥2.5 mg/mL.
  • Storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Typical applications: in vitro and in vivo metabolic enzyme studies.
  • Physical form: light yellow to yellow solid.

Catalog No. 50-002-89026 Supplier Medchemexpress LLC Supplier No. HYN237210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fraxinol is a naturally derived coumarin isolated from Lobelia chinensis and used as a cytochrome P450 inhibitor for research applications. It is supplied as a light yellow solid with high reported purity, documented solubility in DMSO, and recommended storage conditions for both solid and solution forms.

  • Purity: 98.82%.
  • CAS number: 486-28-2.
  • Molecular formula: C11H10O5; molecular weight: 222.19 g·mol⁻¹.
  • Biological target: cytochrome P450 enzymes.
  • Solubility: DMSO 25 mg/mL (in vitro); in vivo formulations yield ≥2.5 mg/mL.
  • Storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Typical applications: in vitro and in vivo metabolic enzyme studies.
  • Physical form: light yellow to yellow solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.