Learn More
Cayman Chemical Fraxnol 10mg
SDP
Catalog No. 502940332
Encompass_Preferred
An Hsp27 and microtubule inhibitor inhibits migration and invasion of MDA-MB-231 cells in vitro in an Hsp27-dependent manner inhibits PKC phosphorylation of Hsp27 induces HCT116 cell cycle arrest in the G2/M phase via formation of the Mad2/p55CDC inhibitory complex and Bax activation of apoptosis inhibits microtubule polymerization in HCT116 cells reduces tumor volume in an HCT116 mouse xenograft model (50-100 mg/kg)
Catalog No. 50-294-0332 Supplier Cayman Chemical Supplier No. 28218 10MG
An Hsp27 and microtubule inhibitor inhibits migration and invasion of MDA-MB-231 cells in vitro in an Hsp27-dependent manner inhibits PKC phosphorylation of Hsp27 induces HCT116 cell cycle arrest in the G2/M phase via formation of the Mad2/p55CDC inhibitory complex and Bax activation of apoptosis inhibits microtubule polymerization in HCT116 cells reduces tumor volume in an HCT116 mouse xenograft model (50-100 mg/kg)