Learn More
Roboz Surgical FRAZIER HOOK 5IN SHP
SDP
Catalog No. 5000574492
Encompass
This Frazier micro dissecting hook is a specialized surgical instrument designed for precision micro-dissection and tissue manipulation. Featuring a sharp tip, this tool is engineered to assist in navigating confined surgical sites during delicate procedures.
  • Sharp tip design for accurate tissue dissection
  • Loop diameter of 2 mm to 3 mm for effective engagement
  • Overall instrument length of 5 in. for balanced handling
Catalog No. 50-005-74492 Supplier Roboz Surgical Supplier No. RS6180
This Frazier micro dissecting hook is a specialized surgical instrument designed for precision micro-dissection and tissue manipulation. Featuring a sharp tip, this tool is engineered to assist in navigating confined surgical sites during delicate procedures.
  • Sharp tip design for accurate tissue dissection
  • Loop diameter of 2 mm to 3 mm for effective engagement
  • Overall instrument length of 5 in. for balanced handling