Learn More
Fisherbrand™ FRC2 Pro Upgradable Phase Contrast Upright with Trinocular Body for Education and Research
GREENER_CHOICE
Catalog No. 03000125 Shop All Fisherbrand Products
Change view
Click to view available options
Magnification Power:
10X, 20X, 40X, 100X
Catalog No. Magnification Power
03-000-125 10X, 20X, 40X, 100X
1 options
Catalog No. 03-000-125
Add to Cart
Add to Cart
FRC2P phase contrast microscope enhances the visibility of transparent, unstained specimens by converting phase shifts in light into contrast.
Widely used in biological research, clinical labs, and education to observe live cells, tissues, and microorganisms without staining. This allows users to study cell behavior, movement, and internal structures in real time. In medical labs, it aids in analyzing urine, blood, and cultured cells. In research, it's essential for live cell imaging and time-lapse studies. Though more complex and expensive than brightfield microscopes, it offers superior contrast for transparent samples, making it a powerful tool for detailed, non-invasive observation.
  • FRC2 Pro Bright Field Microscope
  • Trinocular body
  • 5POS nose
  • 10x, 20x, 40x, and 100x oil PL PH achromat objectives
  • Phase contrast plate set
  • Centering telescope
  • 1.25NA ABBE condenser
  • EvenLIT LED Kohler Illumination
Specifications
Eyepiece 10X/20 mm
Head Style Trinocular
Lighting Type LED
Magnification Power 10X, 20X, 40X, 100X
Model FRC2P-PH1
Objective Types Phase Contrast
Product Type Microscope
Voltage 120 V