Learn More
Fisherbrand™ FRC2 Pro Upgradable Bright Field Upright with Trinocular Body for Education and Research
GREENER_CHOICE
Catalog No. 03000114 Shop All Fisherbrand Products
Change view
Click to view available options
Magnification Power:
4X, 10X, 40X, 100X
Catalog No. Magnification Power
03-000-114 4X, 10X, 40X, 100X
1 options
Catalog No. 03-000-114
Get a Laxco Camera & Accessories with Purchase
View all promotions
Add to Cart
Add to Cart
FRC2P brightfield upright microscope is a simple, cost-effective tool used across medical, educational, research, and industrial fields.
Provides clear visualization of stained or naturally pigmented samples against a bright background. In healthcare, it aids in disease diagnosis; in education, it supports biology teaching; in research, it enables cell and microorganism analysis; and in industry, it ensures quality control of materials. Its ease of use, reliability, and versatility make it ideal for routine lab work and general-purpose microscopy.
  • FRC2 Pro Bright Field Upright Microscope
  • Trinocular body
  • 5-POS nosepiece
  • 4x, 10x, 40x, and 100x oil plan achromat objectives
  • RH mechanical stage
  • 1.25NA ABBE condenser
  • EvenLIT LED Kohler Illumination
Specifications
Eyepiece 10X/20 mm
Head Style Trinocular
Lighting Type LED
Magnification Power 4X, 10X, 40X, 100X
Model FRC2P-BF1
Objective Types Brightfield
Product Type Microscope
Voltage 120 V