Learn More
Med Vet International Bone Reduction Forceps, 8", 2" Ratchet, German Grade, Von Klaus
Catalog No. 503046165
Encompass_Preferred
8" Bone reduction forceps is a German grade surgical instrument. It features a 2" ratchet that allows opening of the jqws to 4" and 8". The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6165 Supplier Med Vet International Supplier No. VK1351149
8" Bone reduction forceps is a German grade surgical instrument. It features a 2" ratchet that allows opening of the jqws to 4" and 8". The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.